Läs mer
meta-Topolin Riboside, TRC
Produktkod. 30012530
Change view
Kvantitet:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012530 50 mg
30012023 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30012530 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT54026550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 110505-76-5
Molekylformel C17H19N5O5
Molekylvikt (g/mol) 373.36
Synonym N-[(3-Hydroxyphenyl)methyl]adenosine, Meta-topolin riboside, meta-Topolin-9-riboside, N-(3-Hydroxybenzyl)adenosine, N6-(3-Hydroxybenzyl)adenosine, N6-(m-Hydroxybenzyl)adenosine, N6-(m-Hydroxybenzyl)adenosine, 6-(3-Hydroxybenzylamino)-9-ß-D-ribofuranosylpurine
IUPAC-namn (2R,3R,4S,5R)-2-(hydroxymethyl)-5-[6-[(3-hydroxyphenyl)methylamino]purin-9-yl]oxolane-3,4-diol
LEDER OC[C@H]1O[C@H]([C@@H](O)[C@H]1O)n2cnc3c(NCc4cccc(O)c4)ncnc23
Specifikationer
Kemiskt namn eller material meta-Topolin Riboside
Kvantitet 50 mg
InChI formel InChI=1S/C17H19N5O5/c23-6-11-13(25)14(26)17(27-11)22-8-21-12-15(19-7-20-16(12)22)18-5-9-2-1-3-10(24)4-9/h1-4,7-8,11,13-14,17,23-26H,5-6H2,(H,18,19,20)/t11-,13+,14+,17-/m1/s1
Formel vikt 373.1386