Läs mer
Metadoxine, TRC
Produktkod. 30483521
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30483521 5 g
1 options
Produktkod. 30483521 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2587655G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Metadoxine
CAS 74536-44-0
Molekylformel C8 H11 N O3 . C5 H7 N O3
Kvantitet 5 g
Synonym L-Proline, 5-oxo-, compd. with 5-hydroxy-6-methyl-3,4-pyridinedimethanol (1:1), 3,4-Pyridinedimethanol, 5-hydroxy-6-methyl-, compd. with 5-oxo-L-proline (1:1), Alcoliv, Metadoxil, Metadoxine, Metasin
InChI formel InChI=1S/C8H11NO3.C5H7NO3/c1-5-8(12)7(4-11)6(3-10)2-9-5;7-4-2-1-3(6-4)5(8)9/h2,10-12H,3-4H2,1H3;3H,1-2H2,(H,6,7)(H,8,9)/t;3-/m.0/s1
LEDER Cc1ncc(CO)c(CO)c1O.OC(=O)[C@@H]2CCC(=O)N2
IUPAC-namn 4,5-bis(hydroxymethyl)-2-methylpyridin-3-ol;(2S)-5-oxopyrrolidine-2-carboxylic acid
Molekylvikt (g/mol) 298.29
Formel vikt 298.1165