Läs mer
Metaflumizone, TRC
Produktkod. 17983945
Change view
Kvantitet:
50 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17983945 100 mg
17942895 50 mg
17914125 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17983945 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM225825100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 139968-49-3
Molekylformel C24 H16 F6 N4 O2
Molekylvikt (g/mol) 506.4
Synonym 2-[2-(4-Cyanophenyl)-1-[3-(trifluoromethyl)phenyl]ethylidene]-N-[4-(trifluoromethoxy)phenyl]hydrazinecarboxamide, Altrevin, Alverde, BAS 320, BAS 32001, BAS 320021, BAS 3201, BAS 320I, BASF 320, Metaflumizone, NNI 0250
IUPAC-namn 1-[[2-(4-cyanophenyl)-1-[3-(trifluoromethyl)phenyl]ethylidene]amino]-3-[4-(trifluoromethoxy)phenyl]urea
LEDER FC(F)(F)Oc1ccc(NC(=O)NN=C(Cc2ccc(cc2)C#N)c3cccc(c3)C(F)(F)F)cc1
Specifikationer
Kemiskt namn eller material Metaflumizone
Kvantitet 100 mg
InChI formel InChI=1S/C24H16F6N4O2/c25-23(26,27)18-3-1-2-17(13-18)21(12-15-4-6-16(14-31)7-5-15)33-34-22(35)32-19-8-10-20(11-9-19)36-24(28,29)30/h1-11,13H,12H2,(H2,32,34,35)
Formel vikt 506.1177