Läs mer
Metahexamide, TRC
Produktkod. 30468130
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30468130 5 mg
1 options
Produktkod. 30468130 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2256005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Metahexamide
CAS 565-33-3
Molekylformel C14 H21 N3 O3 S
Kvantitet 5 mg
Synonym Benzenesulfonamide, 3-amino-N-[(cyclohexylamino)carbonyl]-4-methyl-, Urea, 1-cyclohexyl-3-(4-methylmetanilyl)- (6CI,7CI,8CI), 3-Amino-N-[(cyclohexylamino)carbonyl]-4-methylbenzenesulfonamide, 1-(3-Amino-p-tolylsulfonyl)-3-cyclohexylurea, 3-Amino-4-methylbenzenesulfonylcyclohexylurea, Euglycin, Glyhexylamide, Glyhexylamine isodiane, Melanex, Metahexamide, Methahexamide, Methexamide, N-Cyclohexyl-N'-(3-amino-4-methylbenzenesulfonyl)urea, S 1600, WP 40
InChI formel InChI=1S/C14H21N3O3S/c1-10-7-8-12(9-13(10)15)21(19,20)17-14(18)16-11-5-3-2-4-6-11/h7-9,11H,2-6,15H2,1H3,(H2,16,17,18)
LEDER Cc1ccc(cc1N)S(=O)(=O)NC(=O)NC2CCCCC2
IUPAC-namn 1-(3-amino-4-methylphenyl)sulfonyl-3-cyclohexylurea
Molekylvikt (g/mol) 311.4
Formel vikt 311.1304