Läs mer
Viking Optical™ Metal Case Hygrometer
Brand: Viking Optical™ 7203
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Metal case. Please note that all hair hygrometers require periodic recalibration, at approximately six month intervals.Produkttitel