Learn More
Cole-Parmer™ Metal Manifold, 8 outlets
Expand flow and line capacity with these rugged industrial application manifolds.
Brand:  Cole-Parmer™ 31521-49
Additional Details : Weight : 0.80000kg
Product Code. 11760149
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Ideal for rugged industrial applications.
  • Available in a variety of configurations.
Specifications
Specifications
304 Stainless Steel manifold; 8 outlets; 1/4 in. inlet NPT(F)
15.88 cm
-54°C
2.54 cm
250°F
Stainless Steel (304)
-65°F
1/4 in. inlet NPT(F)
2.54 cm
121°C