Learn More
Cole-Parmer™ Metal Manifold, 8 outlets
Expand flow and line capacity with these rugged industrial application manifolds.
Brand: Cole-Parmer™ 31521-49
Additional Details : Weight : 0.80000kg
Description
- Ideal for rugged industrial applications.
- Available in a variety of configurations.
Specifications
304 Stainless Steel manifold; 8 outlets; 1/4 in. inlet NPT(F) | |
15.88 cm | |
-54°C | |
2.54 cm | |
250°F |
Stainless Steel (304) | |
-65°F | |
1/4 in. inlet NPT(F) | |
2.54 cm | |
121°C |