missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Metal Manifold, 8 outlets

Expand flow and line capacity with these rugged industrial application manifolds.

Brand:  Cole-Parmer™ 31523-19

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11750149

Quantity Price
1 789.00 SEK / Each
10 739.77 SEK / Each
Estimated Shipment
06-07-2021
Add to basket


Description

Description

  • Ideal for rugged industrial applications.
  • Available in a variety of configurations.
Specifications

Specifications

250°F
Brass
Brass Manifold
-54°C
2.54 cm
121°C
15.88 cm
-65°F
1/4 in.
2.54 cm
Videos
SDS
Documents