Beskrivning
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Specifikationer
Specifikationer
| Kemiskt namn eller material | Metalaxyl |
| CAS | 57837-19-1 |
| CAS Max % | ≤ 100% |
| Beilstein | 2947777 |
| Färg | White |
| Smältpunkt | 63.5°C to 72.3°C |
| Kokpunkt | 295.9°C |
| Synonym | N-(2,6-Dimethylphenyl)-N-(methoxyacetyl)-DL-alanine methyl ester |
| Molekylvikt (g/mol) | 279.3 g/mol |
| Kvantitet | 100 mg |