Läs mer
Metalaxyl, TRC
Produktkod. 30476405
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30476405 1 g
1 options
Produktkod. 30476405 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2587401G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Metalaxyl
CAS 57837-19-1
Molekylformel C15 H21 N O4
Kvantitet 1 g
Synonym Alanine, N-(2,6-dimethylphenyl)-N-(methoxyacetyl)-, methyl ester (9CI), DL-Alanine, N-(2,6-dimethylphenyl)-N-(methoxyacetyl)-, methyl ester, (±)-Metalaxyl, Allegiance, Apron, Apron 350FW, Apron 35J, Apron 35WS, Apron FL, Apron SD 35, Apron TL, CG 117, CGA 48988, Fonganil Neu, Himil Gold, IPO-FS, Jiashuangling, Krilaxyl, Metalaxil, Metalaxyl, Metanaxin, Metasyl, Metaxanine, Methyl N-methoxyacetyl-N-(2,6-dimethylphenyl)-DL-alaninate, Ridomil, Ridomil 72WP, Ridomil Vino, Sebring, Subdue
InChI formel InChI=1S/C15H21NO4/c1-10-7-6-8-11(2)14(10)16(13(17)9-19-4)12(3)15(18)20-5/h6-8,12H,9H2,1-5H3
LEDER COCC(=O)N(C(C)C(=O)OC)c1c(C)cccc1C
IUPAC-namn methyl 2-(N-(2-methoxyacetyl)-2,6-dimethylanilino)propanoate
Molekylvikt (g/mol) 279.33
Formel vikt 279.1471