Läs mer
Metam-sodium Hydrate, TRC
Produktkod. 30483763
Change view
Kvantitet:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30483763 1 g
30466288 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30483763 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2258101G
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6734-80-1
Molekylformel C2 H4 N S2 . Na . 2 H2 O
Molekylvikt (g/mol) 165.21
Synonym Carbamodithioic acid, methyl-, monosodium salt, dihydrate (9CI), Carbamic acid, methyldithio-, monosodium salt, dihydrate (8CI), N 869, N-Methylaminomethanethionothiolic acid sodium salt dihydrate, Sodium N-methyldithiocarbamate dihydrate
IUPAC-namn sodium;N-methylcarbamodithioate;dihydrate
LEDER O.O.[Na+].CNC(=S)[S-]
Specifikationer
Kemiskt namn eller material Metam-sodium dihydrate
Kvantitet 1 g
InChI formel InChI=1S/C2H5NS2.Na.2H2O/c1-3-2(4)5;;;/h1H3,(H2,3,4,5);;2*1H2/q;+1;;/p-1
Formel vikt 164.9894