Läs mer
Metanicotine, TRC
Produktkod. 30467691
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30467691 10 mg
30465652 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30467691 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM32125010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 15585-43-0
Molekylformel C10 H14 N2
Molekylvikt (g/mol) 162.23
Synonym 3-Buten-1-amine, N-methyl-4-(3-pyridinyl)-, (3E)-, 3-Buten-1-amine, N-methyl-4-(3-pyridinyl)-, (E)-, Pyridine, 3-[4-(methylamino)-1-butenyl]-, (E)- (8CI), (3E)-N-Methyl-4-(3-pyridinyl)-3-buten-1-amine, (E)-Metanicotine, RJR 2403, Rivanicline, trans-Metanicotine
IUPAC-namn (E)-N-methyl-4-pyridin-3-ylbut-3-en-1-amine
LEDER CNCC\C=C\c1cccnc1
Specifikationer
Kemiskt namn eller material Metanicotine
Kvantitet 10 mg
InChI formel InChI=1S/C10H14N2/c1-11-7-3-2-5-10-6-4-8-12-9-10/h2,4-6,8-9,11H,3,7H2,1H3/b5-2+
Formel vikt 162.1157