Läs mer
Metanilic Acid Sodium Salt (Technical Grade), TRC
Produktkod. 30136589
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136589 500 mg
30134120 50 mg
30007674 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136589 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM321898500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1126-34-7
Molekylformel C6H6NNaO3S
Molekylvikt (g/mol) 195.17
Synonym Benzenesulfonic acid, 3-amino-, sodium salt (1:1) (ACI), Benzenesulfonic acid, 3-amino-, monosodium salt (9CI), Metanilic acid sodium salt (6CI), Metanilic acid, monosodium salt (8CI), 1-Aminobenzene-3-sulfonic acid sodium salt, 3-Aminobenzenesulfonic acid monosodium salt, 3-Aminobenzenesulfonic acid sodium salt, Sodium 1-amino-3-benzenesulfonate, Sodium 3-aminobenzenesulfonate, Sodium m-aminobenzenesulfonate, Sodium metanilate, Aniline-3-sulfonic acid sodium salt
IUPAC-namn sodium;3-aminobenzenesulfonate
LEDER [Na+].Nc1cccc(c1)S(=O)(=O)[O-]
Specifikationer
Kemiskt namn eller material Metanilic Acid Sodium Salt (Technical Grade)
Kvantitet 500 mg
InChI formel InChI=1S/C6H7NO3S.Na/c7-5-2-1-3-6(4-5)11(8,9)10;/h1-4H,7H2,(H,8,9,10);/q;+1/p-1
Formel vikt 194.9966