Läs mer
Metaraminol Bitartrate, TRC
Produktkod. 30475938
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30475938 100 mg
30477908 500 mg
30472874 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30475938 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM225565100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 33402-03-8
Molekylformel C9 H13 N O2 . C4 H6 O6
Molekylvikt (g/mol) 317.29
Synonym Benzenemethanol, α-[(1S)-1-aminoethyl]-3-hydroxy-, (αR)-, (2R,3R)-2,3-dihydroxybutanedioate (1:1), Benzenemethanol, α-(1-aminoethyl)-3-hydroxy-, [R-(R*,S*)]-, [R-(R*,R*)]-2,3-dihydroxybutanedioate (1:1) (salt), Benzenemethanol, α-[(1S)-1-aminoethyl]-3-hydroxy-, (αR)-, (2R,3R)-2,3-dihydroxybutanedioate (1:1) (salt) (9CI), Benzyl alcohol, α-(1-aminoethyl)-m-hydroxy-, (-)-, tartrate (1:1) (salt) (8CI), (-)-Metaraminol bitartrate, Aramine, Aramine (sympathomimetic), Icoral B, Metaradrine bitartrate, Metaraminol bitartrate, Metaraminol hydrogen tartrate, Metaraminol tartrate, Pressorol
IUPAC-namn 3-[(1R,2S)-2-amino-1-hydroxypropyl]phenol;(2R,3R)-2,3-dihydroxybutanedioic acid
LEDER C[C@H](N)[C@H](O)c1cccc(O)c1.O[C@H]([C@@H](O)C(=O)O)C(=O)O
Specifikationer
Kemiskt namn eller material Metaraminol Tartrate
Kvantitet 100 mg
InChI formel InChI=1S/C9H13NO2.C4H6O6/c1-6(10)9(12)7-3-2-4-8(11)5-7;5-1(3(7)8)2(6)4(9)10/h2-6,9,11-12H,10H2,1H3;1-2,5-6H,(H,7,8)(H,9,10)/t6-,9-;1-,2-/m01/s1
Formel vikt 317.1111