Läs mer
Metaxalone, TRC
Produktkod. 30474186
Change view
Kvantitet:
10 mg
25 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30474186 250 mg
30471504 10 mg
30474046 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30474186 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM225850250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1665-48-1
Molekylformel C12 H15 N O3
Molekylvikt (g/mol) 221.25
Synonym 5-[(3,5-Dimethylphenoxy)methyl]-2-oxazolidinone, 2-Oxazolidinone, 5-[(3,5-xylyloxy)methyl]- (6CI,7CI,8CI), 5-[(3,5-Dimethylphenoxy)methyl]-2-oxazolidinone, 5-[(3,5-Xylyloxy)methyl]-2-oxazolidinone, AHR 438, Flexura, Metaxalon, Metaxalone, Metazalone, Metazolone, NSC 170959, Skelaxin, Zorane
IUPAC-namn 5-[(3,5-dimethylphenoxy)methyl]-1,3-oxazolidin-2-one
LEDER Cc1cc(C)cc(OCC2CNC(=O)O2)c1
Specifikationer
Kemiskt namn eller material Metaxalone
Kvantitet 250 mg
InChI formel InChI=1S/C12H15NO3/c1-8-3-9(2)5-10(4-8)15-7-11-6-13-12(14)16-11/h3-5,11H,6-7H2,1-2H3,(H,13,14)
Formel vikt 221.1052