Läs mer
Metazachlor Ethane Sulfonic Acid Sodium Salt, TRC
Produktkod. 17982395
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17982395 50 mg
17914805 25 mg
17974955 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17982395 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM22623350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1231710-68-1
Molekylformel C14H16N3NaO4S
Molekylvikt (g/mol) 345.35
Synonym 2-[(2,6-Dimethylphenyl)(1H-pyrazol-1-ylmethyl)amino]-2-oxoethanesulfonic Acid Sodium Salt, BH 479-8 Sodium Salt;
IUPAC-namn [2-[2,6-dimethyl-N-(pyrazol-1-ylmethyl)anilino]-2-oxo-ethyl]sulfonyloxysodium
LEDER Cc1cccc(C)c1N(Cn2cccn2)C(=O)CS(=O)(=O)O[Na]
Specifikationer
Kemiskt namn eller material Metazachlor Ethane Sulfonic Acid Sodium Salt
Kvantitet 50 mg
InChI formel InChI=1S/C14H17N3O4S.Na/c1-11-5-3-6-12(2)14(11)17(10-16-8-4-7-15-16)13(18)9-22(19,20)21;/h3-8H,9-10H2,1-2H3,(H,19,20,21);/q;+1/p-1
Formel vikt 345.076