Läs mer
Methabenzthiazuron, TRC
Produktkod. 30133094
Change view
Kvantitet:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133094 5 g
17924565 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30133094 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2588805G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 18691-97-9
Molekylformel C10 H11 N3 O S
Molekylvikt (g/mol) 221.28
Synonym Urea, 1-(2-benzothiazolyl)-1,3-dimethyl- (8CI), N-2-Benzothiazolyl-N,N'-dimethylurea, 1,3-Dimethyl-3-(2-benzothiazolyl)urea, 1-(2-Benzothiazolyl)-1,3-dimethylurea, 5633, BAY 5653, Bayer 5653, Bayer 74283, Metabenzthiazuron, Methabenzthiazuron, Methbenzthiazuron, Methibenzuron, Preparation 5633, Tribunil
IUPAC-namn 1-(1,3-benzothiazol-2-yl)-1,3-dimethylurea
LEDER CNC(=O)N(C)c1nc2ccccc2s1
Specifikationer
Kemiskt namn eller material Methabenzthiazuron
Kvantitet 5 g
InChI formel InChI=1S/C10H11N3OS/c1-11-9(14)13(2)10-12-7-5-3-4-6-8(7)15-10/h3-6H,1-2H3,(H,11,14)
Formel vikt 221.0623