Läs mer
Methacetin-13C, TRC
Produktkod. 30012891
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012891 5 mg
30006684 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30012891 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2587725MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 72156-70-8
Molekylformel 13C C8 H11 N O2
Molekylvikt (g/mol) 166.18
Synonym Acetamide, N-[4-(methoxy-13C)phenyl]-, N-[4-(Methoxy-13C)phenyl]acetamide, Methacetin-13C, Methacetin (13C), Methacetin-(methoxy-13C), 4'-(Methoxy-13C)acetanilide, N-Acetyl-4-(methoxy-13C)aniline, N-Acetyl-p-(methoxy-13C)aniline
IUPAC-namn N-(4-(1^{13}C)methoxyphenyl)acetamide
LEDER [13CH3]Oc1ccc(NC(=O)C)cc1
Specifikationer
Kemiskt namn eller material Methacetin-13C
Kvantitet 5 mg
InChI formel InChI=1S/C9H11NO2/c1-7(11)10-8-3-5-9(12-2)6-4-8/h3-6H,1-2H3,(H,10,11)/i2+1
Formel vikt 166.0823