Läs mer
Methacetin, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Methacetin |
| CAS | 51-66-1 |
| Molekylformel | C9 H11 N O2 |
| Kvantitet | 500 mg |
| Synonym | Acetamide, N-(4-methoxyphenyl)-, p-Acetaniside (4CI), p-Acetanisidide (6CI,7CI,8CI), N-(4-Methoxyphenyl)acetamide, 4-(Acetylamino)anisole, 4-Acetamidoanisole, 4-Acetamidophenyl methyl ether, 4-Methoxyacetanilide, 4-Methoxyanilino methyl ketone, 4'-Methoxyacetanilide, Aceto-p-anisidide, Acetyl-p-anisidine, Metacetin, Methacetin, N-(p-Methoxyphenyl)acetamide, N-Acetyl-4-methoxyaniline, N-Acetyl-p-anisidine, N-Acetyl-p-methoxyaniline, NSC 4687, p-Acetanisidine, p-Methoxyacetanilide |
| InChI formel | InChI=1S/C9H11NO2/c1-7(11)10-8-3-5-9(12-2)6-4-8/h3-6H,1-2H3,(H,10,11) |
| LEDER | COc1ccc(NC(=O)C)cc1 |
| IUPAC-namn | N-(4-methoxyphenyl)acetamide |
| Molekylvikt (g/mol) | 165.19 |
| Formel vikt | 165.079 |