missing translation for 'onlineSavingsMsg'
Learn More

Methanesulfinic acid, sodium salt, 85%, Thermo Scientific Chemicals

Catalog Number p-4509458
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
20277-69-4
CH3NaO2S
102.09
LYPGDCWPTHTUDO-UHFFFAOYSA-M
sodium methanesulfinate, sodium methanesulphinate, methanesulfinic acid sodium salt, sodium methylsulfinate, methanesulfinic acid, sodium salt, sodium methane sulfinate, methanesulfinicacidsodiumsalt, methanesulphinic acid sodium salt, methane sulfinic acid sodium salt, meso2na
2733271
sodium;methanesulfinate
CS(=O)[O-].[Na+]
This item is not returnable. View return policy
20277-69-4
CH3NaO2S
102.09
LYPGDCWPTHTUDO-UHFFFAOYSA-M
sodium methanesulfinate, sodium methanesulphinate, methanesulfinic acid sodium salt, sodium methylsulfinate, methanesulfinic acid, sodium salt, sodium methane sulfinate, methanesulfinicacidsodiumsalt, methanesulphinic acid sodium salt, methane sulfinic acid sodium salt, meso2na
2733271
sodium;methanesulfinate
CS(=O)[O-].[Na+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 222.0°C to 226.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 85%
Packaging Glass bottle
Linear Formula CH3SO2Na
Quantity 1 g
Solubility Information Solubility in water: soluble
Formula Weight 102.09
Percent Purity 85%
Physical Form Powder
Chemical Name or Material Methanesulfinic acid, sodium salt
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'