missing translation for 'onlineSavingsMsg'
Learn More
Methyl Alcohol, MP Biomedicals™ Product Code.: 11415366

Methyl Alcohol, MP Biomedicals™

Product Code. p-4185930 Shop All MP Biomedical Products
6 x 1 L, 6 L
Click to view available options
Quantity:
4 x 4 L
6 x 1 L
Unit Size:
16 L
6 L
This item is not returnable. View return policy

Product Code. 11415366

Brand: MP Biomedicals 0230014106

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Methyl Alcohol is used as industrial solvent.

  • Starting material for organic synthesis of formaldehydes, celluloid, textile soap, wood stains, coated fabrics, shatter-proof glass, paper coating, waterproofing formulations, artificial leather, synthetic indigo and other dyes.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Quantity 6 x 1 L
Formula Weight 32.04

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl Alcohol, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.