missing translation for 'onlineSavingsMsg'
Learn More

Methanol, for HPLC, Gradient Grade, ACS, Honeywell Burdick & Jackson™

Click to view available options
Quantity:
7 L
30 L
45 L
395 L
This item is not returnable. View return policy
67-56-1
32.04 g/mol
This item is not returnable. View return policy
67-56-1
32.04 g/mol
CAS Max % 1
Density 0.79 g/cm3 (20°C)
Boiling Point 64°C to 65°C (1,013 hPa)
Color Colorless
Melting Point -98°C
Quantity 395 L
Flash Point 11°C
Chemical Name or Material Methanol