missing translation for 'onlineSavingsMsg'
Läs mer

Methasterone, LoGiCal

Produktkod. 30460901
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30460901 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30460901 Leverantör LoGiCal Leverantörsnummer LGCFOR3331.00

Please to purchase this item. Need a web account? Register with us today!

Denna vara kräver en prisuppdatering innan den beställs.

Begär ett pris för denna vara här.

Denna artikel kan inte returneras. Se returpolicy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material Methasterone
CAS 3381-88-2
Analyt- eller komponentnamn Methasterone
Frakt skick Room Temperature
Molekylformel C21 H34 O2
Certifikat och efterlevnad ISO 9001
Synonym 5α-Androstan-3-one, 17β-hydroxy-2α,17-dimethyl- (6CI,7CI,8CI), 17-Methyldrostanolone, 17β-Hydroxy-2α,17-dimethyl-5α-androstan-3-one, 2α,17α-Dimethyldihydrotestosterone, Methasteron, NSC 40490, Superdrol
InChI formel InChI=1S/C21H34O2/c1-13-12-19(2)14(11-18(13)22)5-6-15-16(19)7-9-20(3)17(15)8-10-21(20,4)23/h13-17,23H,5-12H2,1-4H3/t13-,14+,15-,16+,17+,19+,20+,21+/m1/s1
Lösningstyp Neat
LEDER C[C@@H]1C[C@@]2(C)[C@@H](CC[C@@H]3[C@@H]2CC[C@@]4(C)[C@H]3CC[C@]4(C)O)CC1=O
IUPAC-namn (2R,5S,8R,9S,10S,13S,14S,17S)-17-hydroxy-2,10,13,17-tetramethyl-2,4,5,6,7,8,9,11,12,14,15,16-dodecahydro-1H-cyclopenta[a]phenanthren-3-one
Industry Type Clinical Testing
Molekylvikt (g/mol) 318.49
Kvantitet 10 mg
Formel vikt 318.2559 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.