Läs mer
Methicillin sodium salt, MedChemExpress
Produktkod. 16444517
Encompass_Preferred
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
16444517 50 mg
1 options
Produktkod. 16444517 Leverantör Medchem Express Leverantörsnummer HYB0974
MedChemExpress Methicillin sodium salt (Meticillin sodium) is a β-lactam, semi-synthetic antibiotic related to penicillin antibiotic. Methicillin sodium salt inhibits penicillin-binding proteins involved in the synthesis of peptidoglycan. Methicillin sodium salt inhibits S. aureus with a MIC value of 2.1 μg/ml. Methicillin sodium salt can be used for the research of inflammation.
Methicillin sodium salt (Meticillin sodium) is a β-lactam, semi-synthetic antibiotic related to penicillin antibiotic. Methicillin sodium salt inhibits penicillin-binding proteins involved in the synthesis of peptidoglycan. Methicillin sodium salt inhibits S. aureus with a MIC value of 2.1 μg/ml. Methicillin sodium salt can be used for the research of inflammation.
Specifikationer
Kemiskt namn eller material Methicillin sodium salt
Fysisk form Solid
CAS 132-92-3
Färg White
Kvantitet 50 mg
Anteckningar om renhetsgrad Research
Molekylformel C17H19N2NaO6S
För användning med (applikation) COVID-19-immunoregulation
Synonym Meticillin sodium
Löslighetsinformation DMSO : 125 mg/mL (310.64 mM; Need ultrasonic) ∣H2O : 100 mg/mL (248.51 mM; Need ultrasonic)
LEDER O=C([C@H](N1C2=O)C(C)(S[C@@]1([C@@H]2NC(C3=C(C=CC=C3OC)OC)=O)[H])C)O[Na]
Molekylvikt (g/mol) 402.4
Formel vikt 402.4
Procent renhet 95.0%