missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Beskrivning
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Specifikationer
Specifikationer
| Kemiskt namn eller material | Methiocarb-sulfone |
| CAS | 2179-25-1 |
| CAS Max % | ≤ 100% |
| Beilstein | 2291460 |
| Färg | White |
| Smältpunkt | 158°C to 162°C |
| Synonym | Mercaptodimethursulfon |
| Molekylvikt (g/mol) | 257.31 g/mol |
| Kvantitet | 50 mg |
| Kvalitet | Acaricides and Insecticides |
Produkttitel
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?