Läs mer
Methiocarb Sulfone, TRC
Produktkod. 17950825
Change view
Kvantitet:
250 mg
2.5 g
Förpackningsstorlek:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
17950825 2.5 g 2.5g
17923645 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17950825 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2603402.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2179-25-1
Molekylformel C11 H15 N O4 S
Molekylvikt (g/mol) 257.31
Synonym Carbamic acid, methyl-, 4-(methylsulfonyl)-3,5-xylyl ester (7CI,8CI), Phenol, 3,5-dimethyl-4-(methylsulfonyl)-, methylcarbamate (9CI), 3,5-Xylenol, 4-(methylsulfonyl)-, methylcarbamate (8CI), BAY 41790, Ba 51-084786, Bay 37344 sulfone, Bayer 37344 Sulfone, Mesurol sulfone, Methiocarb sulfone, Methiocarb Sulfone
IUPAC-namn (3,5-dimethyl-4-methylsulfonylphenyl) N-methylcarbamate
LEDER CNC(=O)Oc1cc(C)c(c(C)c1)S(=O)(=O)C
Specifikationer
Kemiskt namn eller material Methiocarb sulfone
Anteckningar om renhetsgrad HPLC
Kvantitet 2.5 g
InChI formel InChI=1S/C11H15NO4S/c1-7-5-9(16-11(13)12-3)6-8(2)10(7)17(4,14)15/h5-6H,1-4H3,(H,12,13)
Formel vikt 257.0722
Procent renhet >95%