Läs mer
Methioprim, TRC
Produktkod. 30479876
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30479876 25 mg
1 options
Produktkod. 30479876 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM26052025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methioprim
CAS 588-36-3
Molekylformel C6H9N3OS
Kvantitet 25 mg
Synonym 4-Amino-2-methylthio-5-pyrimidinemethanol, (4-Amino-2-methanethiopyrimidin-5-yl) methanol, 2-Methylmercapto-4-amino-5-hydroxymethylpyrimidine, 4-Amino-2- methylmercapto-5-pyrimidinemethanol, NSC 3431, NSC 43811
InChI formel InChI=1S/C6H9N3OS/c1-11-6-8-2-4(3-10)5(7)9-6/h2,10H,3H2,1H3,(H2,7,8,9)
LEDER CSc1ncc(CO)c(N)n1
Molekylvikt (g/mol) 171.22