Läs mer
Methiozolin, TRC
Produktkod. 30180123
Change view
Kvantitet:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30180123 5 mg
30180178 25 mg
30180073 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30180123 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM7785055MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 403640-27-7
Molekylformel C17 H17 F2 N O2 S
Molekylvikt (g/mol) 337.3842
Synonym Isoxazole, 5-[[(2,6-difluorophenyl)methoxy]methyl]-4,5-dihydro-5-methyl-3-(3-methyl-2-thienyl)-, 5-[[(2,6-Difluorophenyl)methoxy]methyl]-4,5-dihydro-5-methyl-3-(3-methyl-2-thienyl)isoxazole, Methiozolin
IUPAC-namn 5-[(2,6-difluorophenyl)methoxymethyl]-5-methyl-3-(3-methylthiophen-2-yl)-4H-1,2-oxazole
LEDER Cc1ccsc1C2=NOC(C)(COCc3c(F)cccc3F)C2
Specifikationer
Kemiskt namn eller material Methiozolin
Kvantitet 5 mg
InChI formel InChI=1S/C17H17F2NO2S/c1-11-6-7-23-16(11)15-8-17(2,22-20-15)10-21-9-12-13(18)4-3-5-14(12)19/h3-7H,8-10H2,1-2H3
Formel vikt 337.0948