Läs mer
Methocarbamol, TRC
Produktkod. 30009631
Change view
Kvantitet:
50 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009631 50 mg
30007835 1 g
30007253 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30009631 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM22595050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 532-03-6
Molekylformel C11 H15 N O5
Molekylvikt (g/mol) 241.24
Synonym 1,2-Propanediol, 3-(o-methoxyphenoxy)-, 1-carbamate (8CI), Carbamic acid, 2-hydroxy-3-(o-methoxyphenoxy)propyl ester (6CI,7CI), 2-Hydroxy-3-(o-methoxyphenoxy)propyl carbamate, 3-(2-Methoxyphenoxy)-1-glyceryl carbamate, 3-(o-Methoxyphenoxy)-1,2-propanediol 1-carbamate, 3-(o-Methoxyphenoxy)-2-hydroxypropyl carbamate, AHR 85, Avetil, Delaxin, Etroflex, Glycerylguaiacolate carbamate, Guaiacol glyceryl ether carbamate, Guaiphenesin carbamate, Lumirelax, Methocarbamol, Miolaxene, NSC 170960, Neuraxin, Reflexyn, Relestrid, Robamol, Robaxin, Robinax, Surquetil, Traumacut, Tresortil, guaiamar carbamate, racemic-Methocarbamol
IUPAC-namn [2-hydroxy-3-(2-methoxyphenoxy)propyl] carbamate
LEDER COc1ccccc1OCC(O)COC(=O)N
Specifikationer
Kemiskt namn eller material Methocarbamol
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C11H15NO5/c1-15-9-4-2-3-5-10(9)16-6-8(13)7-17-11(12)14/h2-5,8,13H,6-7H2,1H3,(H2,12,14)
Formel vikt 241.095
Procent renhet >95%