missing translation for 'onlineSavingsMsg'
Läs mer

Methoprene, TRC

Produktkod. 30180350
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30180350 250 mg
30180314 100 mg
30179848 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30180350 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM260715250MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 40596-69-8
Molekylformel C19 H34 O3
Molekylvikt (g/mol) 310.47
Synonym 2,4-Dodecadienoic acid, 11-methoxy-3,7,11-trimethyl-, 1-methylethyl ester, (E,E)-, (2E,4E)-3,7,11-Trimethyl-11-methoxydodeca-2,4-dienoic acid ethyl ester, Altosid, Altosid Liquid Larvicide, Altosid SR 10, Altosid XR, Apex, Apex (pesticide), Apex 5E, Apex 600E, Bioprene BM Fire Ant Killer Bait, Dianex, ENT 70460, Isopropyl (2E,4E)-11-methoxy-3,7,11-trimethyl-2,4-dodecadienoate, Juvemon, Juvenmon, Kabat, Manina, Manta, Manta (hormone), Methoprene, Methoprene S, Metoprag 20CE, SB-515, Starbar Inhibitor, ZPA 1019, ZR 515, dl-Isopropyl 11-methoxy-3,7,11-trimethyl-trans-trans-2,4-dodecadienoate
IUPAC-namn propan-2-yl (2E,4E)-11-methoxy-3,7,11-trimethyldodeca-2,4-dienoate
LEDER COC(C)(C)CCCC(C)C\C=C\C(=C\C(=O)OC(C)C)\C

Specifikationer

Kemiskt namn eller material Methoprene
Kvantitet 250 mg
InChI formel InChI=1S/C19H34O3/c1-15(2)22-18(20)14-17(4)11-8-10-16(3)12-9-13-19(5,6)21-7/h8,11,14-16H,9-10,12-13H2,1-7H3/b11-8+,17-14+
Formel vikt 310.2508
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.