Läs mer
Methotrexate Dimethyl Ester, TRC
Produktkod. 30137748
Change view
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137748 250 mg
30136121 500 mg
17963645 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137748 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM260685250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 34378-65-9
Molekylformel C22 H26 N8 O5
Molekylvikt (g/mol) 482.49
Synonym L-Glutamic acid, N-[4-[[(2,4-diamino-6-pteridinyl)methyl]methylamino]benzoyl]-, 1,5-dimethyl ester, Glutamic acid, N-[p-[[(2,4-diamino-6-pteridinyl)methyl]methylamino]benzoyl]-, dimethyl ester, L-(+)- (8CI), L-Glutamic acid, N-[4-[[(2,4-diamino-6-pteridinyl)methyl]methylamino]benzoyl]-, dimethyl ester (9CI), Dimethyl methotrexate, Methotrexate dimethyl ester, Methotrexate Imp. J (EP)
IUPAC-namn dimethyl (2S)-2-[[4-[(2,4-diaminopteridin-6-yl)methyl-methylamino]benzoyl]amino]pentanedioate
LEDER COC(=O)CC[C@H](NC(=O)c1ccc(cc1)N(C)Cc2cnc3nc(N)nc(N)c3n2)C(=O)OC
Specifikationer
Kemiskt namn eller material Methotrexate Dimethyl Ester
Anteckningar om renhetsgrad HPLC
InChI formel InChI=1S/C22H26N8O5/c1-30(11-13-10-25-19-17(26-13)18(23)28-22(24)29-19)14-6-4-12(5-7-14)20(32)27-15(21(33)35-3)8-9-16(31)34-2/h4-7,10,15H,8-9,11H2,1-3H3,(H,27,32)(H4,23,24,25,28,29)/t15-/m0/s1
Kvantitet 250 mg
Formel vikt 482.2026
Procent renhet >95%