Läs mer
Methoxamine Hydrochloride (Mixture of Diastereomers), TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Methoxamine Hydrochloride |
| CAS | 61-16-5 |
| Molekylformel | C11 H17 N O3 . Cl H |
| Kvantitet | 50 mg |
| Synonym | Methoxamine hydrochloride, 2-Amino-1-(2,5-dimethoxyphenyl)-1-propanol hydrochloride, 2-Amino-1-hydroxy-1-(2,5-dimethoxyphenyl)propane hydrochloride, Beta-hydroxy-beta-(2,5-dimethoxyphenol)-isopropylamine hydrochloride, Pressomin hydrochloride, Vasoxine, Vasoxine hydrochloride, Vasoxyl, Vasoxyl hydrochloride, Vasylox, Benzenemethanol, alpha-(1-aminoethyl)-2,5-dimethoxy-, hydrochloride, Benzyl alcohol, alpha-(1-aminoethyl)-2,5-dimethoxy-, hydrochloride, alpha-(alpha-Aminoethyl)-2,5-dimethoxybenzyl alcohol hydrochloride, beta-Hydroxy-beta-(2,5-dimethoxyphenyl)isopropylamine hydrochloride |
| InChI formel | InChI=1S/C11H17NO3.ClH/c1-7(12)11(13)9-6-8(14-2)4-5-10(9)15-3;/h4-7,11,13H,12H2,1-3H3;1H |
| LEDER | Cl.COc1ccc(OC)c(c1)C(O)C(C)N |
| IUPAC-namn | 2-amino-1-(2,5-dimethoxyphenyl)propan-1-ol;hydrochloride |
| Molekylvikt (g/mol) | 247.72 |
| Formel vikt | 247.0975 |