Läs mer
Methoxy Adrenaline Hydrochloride, TRC
Produktkod. 30138517
Change view
Kvantitet:
250 mg
2.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138517 2.5 g
30007610 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30138517 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2608752.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 74571-90-7
Molekylformel C10H16ClNO3
Molekylvikt (g/mol) 233.69
Synonym 4-[1-Methoxy-2-(methylamino)ethyl]-1,2-benzenediol Hydrochloride, 4-[1-Methoxy-2-(methylamino)ethyl]pyrocatechol Hydrochloride, 4-(α-Methoxy-β-methylaminoethyl)pyrocatechol Hydrochloride
IUPAC-namn 4-[1-methoxy-2-(methylamino)ethyl]benzene-1,2-diol;hydrochloride
LEDER OC1=C(O)C=C(C(OC)CNC)C=C1.Cl
Specifikationer
Kemiskt namn eller material Methoxy Adrenaline Hydrochloride
Anteckningar om renhetsgrad HPLC
Kvantitet 2.5 g
InChI formel InChI=1S/C10H15NO3.ClH/c1-11-6-10(14-2)7-3-4-8(12)9(13)5-7;/h3-5,10-13H,6H2,1-2H3;1H
Formel vikt 233.0819
Procent renhet >95%