Learn More
Methoxy(cyclooctadiene)iridium(I) dimer, Ir nominally 58%, Thermo Scientific Chemicals
Catalog Number p-4781997
Click to view available options
Quantity:
1 g
5 g
12148-71-9
C18H30Ir2O2
662.87
MFCD08459360
SGBSCFWMCJNCRM-MIXQCLKLSA-N
bis 1,5-cyclooctadiene di-mu-methoxydiiridium i, 1,5-cyclooctadiene methoxy iridium i dimer
124202702
(1Z,5Z)-cycloocta-1,5-diene;iridium;methanol
CO[Ir+].CO[Ir+].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1
12148-71-9
C18H30Ir2O2
662.87
MFCD08459360
SGBSCFWMCJNCRM-MIXQCLKLSA-N
bis 1,5-cyclooctadiene di-mu-methoxydiiridium i, 1,5-cyclooctadiene methoxy iridium i dimer
124202702
(1Z,5Z)-cycloocta-1,5-diene;iridium;methanol
CO[Ir+].CO[Ir+].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1
Light, Air & Moisture Sensitive
Methoxy(cyclooctadiene)iridium(I) dimer is used as a catalyst in the preparation of heteroaryl fused indole ring systems and borylation and Suzuki-miyaura coupling. Further, it is used in tetraborylation reactions, ortho-silylation of aryl ketone, benzaldehyde and benzyl alcohol derivatives through C-H activation. The most useful application is highly regio and enantio selection asymmetric hydroboration.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Color Yellow
Assay Percent Range Ir nominally 58%
Quantity 1 g
Sensitivity Air, light, and moisture sensitive
Formula Weight 662.88
Concentration or Composition (by Analyte or Components) Ir nominally 58%
Physical Form Powder
Chemical Name or Material Methoxy(cyclooctadiene)iridium(I) dimer