missing translation for 'onlineSavingsMsg'
Learn More

Methoxyacetic anhydride, 95%, Thermo Scientific Chemicals

Catalog Number p-7055855
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
19500-95-9
C6H10O5
162.14
MFCD11976032
PEHFQQWAINXOQG-UHFFFAOYSA-N
methoxyacetic anhydride, methoxyacetic acid anhydride, acetic acid, methoxy-, anhydride, 2-methoxyacetic anhydride, acetic acid,2-methoxy-, anhydride, di methoxyacetic anhydride, methoxy acetic acid anhydride, methoxy-acetic acid anhydride, 2-methoxyacetyl 2-methoxyacetate, methoxyacetic anhydride?intermediate of 5-aza-2'-deoxycytidine
88095
(2-methoxyacetyl) 2-methoxyacetate
COCC(=O)OC(=O)COC
This item is not returnable. View return policy
19500-95-9
C6H10O5
162.14
MFCD11976032
PEHFQQWAINXOQG-UHFFFAOYSA-N
methoxyacetic anhydride, methoxyacetic acid anhydride, acetic acid, methoxy-, anhydride, 2-methoxyacetic anhydride, acetic acid,2-methoxy-, anhydride, di methoxyacetic anhydride, methoxy acetic acid anhydride, methoxy-acetic acid anhydride, 2-methoxyacetyl 2-methoxyacetate, methoxyacetic anhydride?intermediate of 5-aza-2'-deoxycytidine
88095
(2-methoxyacetyl) 2-methoxyacetate
COCC(=O)OC(=O)COC

Methoxyacetic anhydride is used as the intermediate of 5-Aza-2'-deoxycytidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methoxyacetic anhydride is used as the intermediate of 5-Aza-2′-deoxycytidine.

Solubility
Very Soluble in water.(193 g/L) (25°C)

Notes
Moisture sensitive. Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Color Colorless
Linear Formula (CH3OCH2CO)2O
Quantity 25 g
UN Number UN3265
Beilstein 1771552
Sensitivity Moisture sensitive
Solubility Information Very Soluble in water.(193g/L) (25°C)
Formula Weight 162.14
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Methoxyacetic anhydride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'