missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Methoxycarbonylmethyl)triphenylphosphonium chloride, 97%, Thermo Scientific™
Click to view available options
Quantity:
10 g
50 g
250 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is employed as intermediate for pharmaceutical.
Solubility
Very Soluble in water (100 g/L) (25°C).
Notes
Hygroscopic, store away from moisture. Store in a tightly closed container in a cool, dry, well-ventilated area. Store away from oxidizing agent.
It is employed as intermediate for pharmaceutical.
Solubility
Very Soluble in water (100 g/L) (25°C).
Notes
Hygroscopic, store away from moisture. Store in a tightly closed container in a cool, dry, well-ventilated area. Store away from oxidizing agent.
Specifications
Melting Point | ∼162°C (decomposition) |
Quantity | 10 g |
Beilstein | 4172737 |
Solubility Information | Very Soluble in water (100g/L) (25°C). |
IUPAC Name | (2-methoxy-2-oxoethyl)-triphenylphosphanium;chloride |
Formula Weight | 370.82 |
Percent Purity | 97% |
Chemical Name or Material | (Methoxycarbonylmethyl)triphenylphosphonium chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'