missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Metaxifluran, TRC
Specifications
Specifications
| Kemiskt namn eller material | Methoxyflurane |
| CAS | 76-38-0 |
| Anteckningar om renhetsgrad | GC |
| Molekylformel | C3 H4 Cl2 F2 O |
| Kvantitet | 1 g |
| Synonym | Dichloro-2,2-difluoro-2-methoxyethane, 2,2-Dichloro-1,1-difluoro-1-methoxyethane, 2,2-Dichloro-1,1-difluoroethyl Methyl Ether, Analgizer, Anecotan, DA 759, Inhalan, Methoflurane, Methoxane, Methoxyfluran, Methoxyflurane, Methyl 1,1-difluoro-2,2-dichloroethyl ether, Metofane, Metoxfluran, Metoxifluran, NSC 110432, Penthrane, Pentran, Pentrane |
| InChI formel | InChI=1S/C3H4Cl2F2O/c1-8-3(6,7)2(4)5/h2H,1H3 |
| LEDER | COC(F)(F)C(Cl)Cl |
| IUPAC-namn | 2,2-diklor-1,1-difluor-1-metoxyetan |
| Molekylvikt (g/mol) | 164.97 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?