missing translation for 'onlineSavingsMsg'
Learn More

(Methoxymethyl)triphenylphosphonium chloride, 98%, Thermo Scientific Chemicals

Catalog Number p-157353
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
4009-98-7
C20H20ClOP
342.80
MFCD00011800
SJFNDMHZXCUXSA-UHFFFAOYSA-M
methoxymethyl triphenylphosphonium chloride, methoxymethyl triphenylphosphanium chloride, phosphonium, methoxymethyl triphenyl-, chloride, methoxymethyltriphenylphosphonium chloride, methoxymethyl-triphenylphosphonium chloride, methoxymethyl-triphenyl-phosphonium chloride, triphenyl methoxymethyl phosphonium chloride, methoxy methyl triphenyl phosphonium chloride
2723798
methoxymethyl(triphenyl)phosphanium;chloride
[Cl-].COC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
4009-98-7
C20H20ClOP
342.80
MFCD00011800
SJFNDMHZXCUXSA-UHFFFAOYSA-M
methoxymethyl triphenylphosphonium chloride, methoxymethyl triphenylphosphanium chloride, phosphonium, methoxymethyl triphenyl-, chloride, methoxymethyltriphenylphosphonium chloride, methoxymethyl-triphenylphosphonium chloride, methoxymethyl-triphenyl-phosphonium chloride, triphenyl methoxymethyl phosphonium chloride, methoxy methyl triphenyl phosphonium chloride
2723798
methoxymethyl(triphenyl)phosphanium;chloride
[Cl-].COC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 195.0°C to 197.0°C
Color White
Boiling Point 413.0°C
Flash Point >250°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3OCH2P(C6H5)3Cl
Quantity 25 g
Solubility Information Solubility in water: decomposes
Formula Weight 342.79
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material (Methoxymethyl)triphenylphosphonium chloride, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'