Läs mer
Methyclothiazide, TRC
Produktkod. 30477236
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30477236 50 mg
1 options
Produktkod. 30477236 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM26015550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyclothiazide
CAS 135-07-9
Molekylformel C9 H11 Cl2 N3 O4 S2
Kvantitet 50 mg
Synonym Methyclothiazide, Methyclothiazid, 6-chloro-3-(chloromethyl)-3,4-dihydro-2-methyl-2H-1,2,4-benzothiadiazine-7-sulfonamide-1,1-dioxide, 6-Chloro-3-chloromethyl-3,4-dihydro-2-methyl-7-sulfamoyl-1,2,4-benzothiadiazine 1,1-dioxide, Aquatensen, Ciba 7272-Su, Duretic, Enduron, Enduronum, Eutron, Methyclothiazid, Methyclothiazide, Methycyclothiazide, Methylcyclothiazide, NSC 110431, Naturon
InChI formel InChI=1S/C9H11Cl2N3O4S2/c1-14-9(4-10)13-6-2-5(11)7(19(12,15)16)3-8(6)20(14,17)18/h2-3,9,13H,4H2,1H3,(H2,12,15,16)
LEDER CN1C(CCl)Nc2cc(Cl)c(cc2S1(=O)=O)S(=O)(=O)N
IUPAC-namn 6-chloro-3-(chloromethyl)-2-methyl-1,1-dioxo-3,4-dihydro-1λ^{6},2,4-benzothiadiazine-7-sulfonamide
Molekylvikt (g/mol) 360.24
Formel vikt 358.9568