Läs mer
Methyl 1-(2,4-Difluorophenyl)-3-phenyl-1H-pyrazole-5-carboxylate, TRC
Produktkod. 30472230
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30472230 10 mg
1 options
Produktkod. 30472230 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM33377310MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 1-(2,4-Difluorophenyl)-3-phenyl-1H-pyrazole-5-carboxylate
CAS 1202030-37-2
Molekylformel C17 H12 F2 N2 O2
Kvantitet 10 mg
Synonym 1H-Pyrazole-5-carboxylic acid, 1-(2,4-difluorophenyl)-3-phenyl-, methyl ester (ACI), Methyl 1-(2,4-difluorophenyl)-3-phenyl-1H-pyrazole-5-carboxylate (ACI)
InChI formel InChI=1S/C17H12F2N2O2/c1-23-17(22)16-10-14(11-5-3-2-4-6-11)20-21(16)15-8-7-12(18)9-13(15)19/h2-10H,1H3
LEDER COC(=O)c1cc(nn1c2ccc(F)cc2F)c3ccccc3
IUPAC-namn methyl 2-(2,4-difluorophenyl)-5-phenyl-pyrazole-3-carboxylate
Molekylvikt (g/mol) 314.29
Formel vikt 314.0867