missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methyl 1-aminocyclobutanecarboxylate hydrochloride, 95%

Brand:  Alfa Aesar™ H64701.06

Additional Details : CAS Number : 92398-47-5 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15449086

  • 7626.42 SEK / 5g


Estimated Shipment date
from Supplier 12-11-2018
Add to basket

Description

Description

Applications
Used as a primary and secondary intermediate for pharmaceutical and organic synthesis.

Solubility
Slightly soluble in water.

Notes
Air Sensitive. Store away form air. Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

215597-35-6
129.159
SGXOPTPGZBLQAY-UHFFFAOYSA-N
1502033
COC(=O)C1(CCC1)N
C6H11NO2
MFCD04115288
methyl 1-aminocyclobutanecarboxylate, 1-amino-cyclobutanecarboxylic acid methyl ester, 1-aminocyclobutanecarboxylic acid methyl ester, cyclobutanecarboxylic acid, 1-amino-, methyl ester, 1-amino-cyclobutanecarboxylicacidmethylester, methyl 1-amino-cyclobutane-1-carboxylate, 1-aminocyclobutane-1-carboxylic acid methyl ester, cyclobutanecarboxylicacid, 1-amino-, methyl ester
methyl 1-aminocyclobutane-1-carboxylate

Specifications

Methyl 1-aminocyclobutanecarboxylate hydrochloride
5g
C6H11NO2
5615121
SGXOPTPGZBLQAY-UHFFFAOYSA-N
methyl 1-aminocyclobutane-1-carboxylate
1502033
Solid
Air Sensitive
215597-35-6
MFCD04115288
methyl 1-aminocyclobutanecarboxylate, 1-amino-cyclobutanecarboxylic acid methyl ester, 1-aminocyclobutanecarboxylic acid methyl ester, cyclobutanecarboxylic acid, 1-amino-, methyl ester, 1-amino-cyclobutanecarboxylicacidmethylester, methyl 1-amino-cyclobutane-1-carboxylate, 1-aminocyclobutane-1-carboxylic acid methyl ester, cyclobutanecarboxylicacid, 1-amino-, methyl ester
COC(=O)C1(CCC1)N
129.159
165.62
95%
Videos
SDS