Läs mer
Methyl 1-naphthylacetate, 97%, Thermo Scientific™
Produktkod. 11414484
Change view
Kvantitet:
5 g
25 g
100 g
Förpackningsstorlek:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11414484 5 g 5g
11424484 25 g 25g
11434484 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 11414484 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer A13126.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Detta Thermo Scientific varumärkesprodukt var ursprungligen en del av Alfa Aesar produktportfölj. Viss dokumentation och etikettinformation kan referera till det äldre varumärket. Originalet Alfa Aesar produkt-/artikelkod eller SKU-referens har inte ändrats som en del av varumärkesövergången till Thermo Scientific .
Applications
Methyl 1-naphthylacetate, is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Sparingly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2876-78-0
Molekylformel C13H12O2
Molekylvikt (g/mol) 200.237
MDL-nummer MFCD00004045
InChI-nyckel YGGXZTQSGNFKPJ-UHFFFAOYSA-N
Synonym methyl 1-naphthaleneacetate, methyl 1-naphthylacetate, hormonit, kartofin, methyl 2-naphthalen-1-yl acetate, 1-naphthaleneacetic acid methyl ester, m 1 growth regulator, 1-naphthaleneacetic acid, methyl ester, methyl alpha-naphthylacetate, methyl naphthaleneacetic acid
PubChem CID 17891
IUPAC-namn methyl 2-naphthalen-1-ylacetate
LEDER COC(=O)CC1=CC=CC2=CC=CC=C21
Specifikationer
Kemiskt namn eller material Methyl 1-naphthylacetate
Densitet 1.144
Kokpunkt 155°C to 157°C (9 mmHg)
Flampunkt >110°C (230°F)
Brytningsindex 1.598
Kvantitet 5 g
Beilstein 1956293
Löslighetsinformation Sparingly soluble in water.
Formel vikt 200.24
Procent renhet 97%