Läs mer
Methyl 10-Undecenoate(10-Undecenoic Acid Methyl Ester), TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | 10-Undecenoic acid-methyl ester |
| CAS | 111-81-9 |
| Molekylformel | C12 H22 O2 |
| Kvantitet | 100 mg |
| Synonym | 10-Undecenoic acid-methyl ester, 10-Undecenoic acid','methyl ester (7CI','8CI','9CI','ACI), 10-Hendecenoic acid methyl ester, Maskod MNS 01-10, Methyl 10-undecenoate, Methyl 10-undecylenate, Methyl undec-10-enoate, Methyl undec-10-enylate, Methyl undecenoate, NSC 1273, Undecylenic acid methyl ester |
| InChI formel | InChI=1S/C12H22O2/c1-3-4-5-6-7-8-9-10-11-12(13)14-2/h3H,1,4-11H2,2H3 |
| LEDER | COC(=O)CCCCCCCCC=C |
| IUPAC-namn | methyl undec-10-enoate |
| Molekylvikt (g/mol) | 198.3 |
| Formel vikt | 198.162 |