Läs mer
Methyl 11-Bromoundecanoate, TRC
Produktkod. 30479646
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30479646 500 mg
1 options
Produktkod. 30479646 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM296585500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 11-Bromoundecanoate
CAS 6287-90-7
Molekylformel C12H23O2Br
Kvantitet 500 mg
Synonym 11-bromo-Undecanoic acid methyl ester, 11-bromo-Hendecanoic acid methyl ester, 11-Bromoundecanoic acid methyl ester, NSC 12015
InChI formel InChI=1S/C12H23BrO2/c1-15-12(14)10-8-6-4-2-3-5-7-9-11-13/h2-11H2,1H3
LEDER COC(=O)CCCCCCCCCCBr
IUPAC-namn methyl 11-bromoundecanoate
Molekylvikt (g/mol) 279.21
Formel vikt 278.0881