Läs mer
Methyl 12-Hydroxystearate, TRC
Produktkod. 30482124
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30482124 1 g
1 options
Produktkod. 30482124 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2930501G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 12-hydroxystearate
CAS 141-23-1
Molekylformel C19 H38 O3
Kvantitet 1 g
Synonym 12-Hydroxyoctadecanoic acid methyl ester, Methyl 12-hydroxyoctadecanoate, Methyl 12-hydroxystearate, 12-Hydroxyoctadecanoic acid, methyl ester
InChI formel InChI=1S/C19H38O3/c1-3-4-5-12-15-18(20)16-13-10-8-6-7-9-11-14-17-19(21)22-2/h18,20H,3-17H2,1-2H3
LEDER CCCCCCC(O)CCCCCCCCCCC(=O)OC
IUPAC-namn methyl 12-hydroxyoctadecanoate
Molekylvikt (g/mol) 314.5
Formel vikt 314.2821