Läs mer
Methyl 18-Methyleicosanoate, TRC
Produktkod. 30472965
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30472965 500 mg
1 options
Produktkod. 30472965 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM764120500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 18-Methyleicosanoate
CAS 65301-92-0
Molekylformel C22H44O2
Kvantitet 500 mg
Synonym 18-Methyleicosanoic Acid','Methyl Ester
InChI formel InChI=1S/C22H44O2/c1-4-21(2)19-17-15-13-11-9-7-5-6-8-10-12-14-16-18-20-22(23)24-3/h21H,4-20H2,1-3H3
LEDER O=C(OC)CCCCCCCCCCCCCCCCC(C)CC
Molekylvikt (g/mol) 340.58