Läs mer
Methyl 2-(1-Hexyn-1-yl)benzoate, TRC
Produktkod. 30476089
Change view
Kvantitet:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30476089 50 mg
30480039 250 mg
30469259 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30476089 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM33510550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 462637-40-7
Molekylformel C14H16O2
Molekylvikt (g/mol) 216.28
Synonym 1-(2-Methoxycarbonylphenyl)-1-hexyne, 2-(1-Hexynyl)-benzoic Acid Methyl Ester
IUPAC-namn methyl 2-(hex-1-yn-1-yl)benzoate
LEDER ClCC1=CC=C(SC2=CC=CC=C2)C=C1
Specifikationer
Kemiskt namn eller material Methyl 2-(1-Hexyn-1-yl)benzoate
Kvantitet 50 mg
InChI formel InChI=1S/C14H16O2/c1-3-4-5-6-9-12-10-7-8-11-13(12)14(15)16-2/h7-8,10-11H,3-5H2,1-2H3
Formel vikt 216.12