Läs mer
methyl 2-(2-formylphenyl)acetate, TRC
Produktkod. 30476374
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30476374 50 mg
30471471 100 mg
30467548 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30476374 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM62390550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 63969-83-5
Molekylformel C10H10O3
Molekylvikt (g/mol) 178.18
IUPAC-namn methyl 2-(2-formylphenyl)acetate
LEDER COC(=O)Cc1ccccc1C=O
Specifikationer
Kemiskt namn eller material methyl 2-(2-formylphenyl)acetate
Kvantitet 50 mg
InChI formel InChI=1S/C10H10O3/c1-13-10(12)6-8-4-2-3-5-9(8)7-11/h2-5,7H,6H2,1H3
Formel vikt 178.063