Läs mer
Methyl 2,3,4-Tri-O-benzyl-Beta-D-glucuronic Acid, Benzyl Ester, TRC
Produktkod. 30481959
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30481959 25 mg
1 options
Produktkod. 30481959 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM33082525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 2,3,4-Tri-O-benzyl-beta-D-glucuronic Acid, Benzyl Ester
CAS 4356-83-6
Molekylformel C35H36O7
Kvantitet 25 mg
Synonym Methyl 2,3,4-Tri-O-Benzyl-B-D-Glucuronic Acid, Benzyl Ester
InChI formel InChI=1S/C35H36O7/c1-37-35-33(40-24-28-18-10-4-11-19-28)31(39-23-27-16-8-3-9-17-27)30(38-22-26-14-6-2-7-15-26)32(42-35)34(36)41-25-29-20-12-5-13-21-29/h2-21,30-33,35H,22-25H2,1H3/t30-,31-,32-,33+,35+/m0/s1
LEDER CO[C@@H]1O[C@@H]([C@@H](OCc2ccccc2)[C@H](OCc3ccccc3)[C@H]1OCc4ccccc4)C(=O)OCc5ccccc5
Molekylvikt (g/mol) 568.66