missing translation for 'onlineSavingsMsg'
Läs mer

Methyl 2,3-dichlorobenzoate, TRC

Product Code. 30465201
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Kvantitet:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30465201 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30465201 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCM3014101G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Methyl 2,3-dichlorobenzoate
CAS 2905-54-6
Molekylformel C8H6Cl2O2
Kvantitet 1 g
InChI formel InChI=1S/C8H6Cl2O2/c1-12-8(11)5-3-2-4-6(9)7(5)10/h2-4H,1H3
LEDER COC(=O)C1=C(C(=CC=C1)Cl)Cl
IUPAC-namn methyl 2,3-dichlorobenzoate
Molekylvikt (g/mol) 205.04
Formel vikt 203.9745
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.