Läs mer
Methyl 2,3-Dichlorotrifluoropropionate, TRC
Produktkod. 30477279
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30477279 500 mg
30469253 50 mg
30469549 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30477279 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM696180500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 16329-89-8
Molekylformel C4H3O2F3Cl2
Molekylvikt (g/mol) 210.96
Synonym Methyl 2,3-Dichloro-2,3,3-trifluoropropanoate, Methyl 2,3-Dichloro-2,3,3-trifluoropropionate, Methyl 2,3-Dichlorotrifluoropropionate
IUPAC-namn methyl 2,3-dichloro-2,3,3-trifluoropropanoate
LEDER COC(=O)C(F)(Cl)C(F)(F)Cl
Specifikationer
Kemiskt namn eller material Methyl 2,3-Dichlorotrifluoropropionate
Kvantitet 500 mg
InChI formel InChI=1S/C4H3Cl2F3O2/c1-11-2(10)3(5,7)4(6,8)9/h1H3
Formel vikt 209.9462